Reagent alcohol, anhydrous ACS for organic synthesis, for preparative liquid chromatography (denatured with methanol and isopropanol), Burdick & Jackson™
Catalog # BJAALP090-4
Supplier: BURDICK JACKSON
CAS Number:
Specifications
- Pk:16 L
- Seq:5254
Specifications
Specification Test Results
- Assay (Ethanol + Methanol):94 - 96 %
- Assay (Isopropyl Alcohol):4 - 6 %
- Color (APHA):10 max.
- Residue after Evaporation:0.001 % max.
- Water by Karl Fischer Titration:0.5 % max.
Specification Test Results
About this item
:
Boiling Pt:78,32 °C (main component)
Melting Pt:–114,1 °C (main component)
Density:0,7890 g/cm³ (20 °C)
Flash Pt:15 °C (main component)
Storage Temperature:Ambient
EINECS:200-578-6
UN:1987
ADR:3,II
:GHS07
:GHS08
:GHS02
CAS Number:64-17-5
MDL Number:MFCD00003568