

Dichloromethane ≥99.9% stabilised, B&J Brand™ for HPLC, for pesticide residue analysis, for spectrophotometry, Burdick & Jackson™
Catalog # BJAA299-4
Supplier: BURDICK JACKSON
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:16 L
- Seq:5254
Specifications
Specification Test Results
- Acidity (as HCl):1 mg/L max.
- Chloride:10 mg/L max.
- Electron Capture GC:10 ng/L max.
- Flame Ionization GC (as nonane):5 ppb max.
- GC Analysis:99.9 % min.
- Refractive Index (20ºC):1.4236 - 1.4246
- Residue:1 mg/L max.
- UV Cutoff:233 nm max.
- Water by Karl Fischer Titration:0.01 % max.
Specification Test Results
About this item
:
:84.93 g/mol
Merck Index:12,06140
Boiling Pt:40 °C
Melting Pt:–95 °C
Density:1,33 g/cm³
Flash Pt: Does not flash
EINECS:200-838-9
UN:1593
ADR:6.1,III
:Stabilised with amylene
:GHS07
:GHS08
Storage Temperature:Ambient
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881