
Bis(8-hydroxyquinolinium)sulphate monohydrate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:207386-91-2
:
Formula:C₁₈H₁₆N₂O₆S·1H₂O
MDL Number:MFCD00149610
:388.4 g/mol
Melting Pt:176…179 °C
EINECS:205-137-1
:GHS07
Storage Temperature:Ambient