


Acetonitrile ≥99.9% ACS, meets analytical specification of USP for HPLC, for spectrophotometry
Catalog # BJAAAH015-4
Supplier: BURDICK JACKSON
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:16 L
- Seq:5254
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
- Pk Info:4× 4 L
Specifications
Specification Test Results
- Color (APHA):10 max.
- Water by Karl Fisher Titration:0.05 % max.
- GC Analysis:99.9 % min.
- HPLC Gradient @ 254 nm:0.005 AU max.
- Refractive Index (20ºC):1.3434 - 1.3446
- Residue:3 mg/L max.
- Titrable Acid:8 µeq/g max.
- Titrable Base:0.6 µeq/g max.
- UV Absorbance @ 200 nm:0.07 AU max.
- UV Cutoff:190 nm max.
Specification Test Results
About this item
:
:41.05 g/mol
Merck Index:13,00071
Boiling Pt:82 °C (1,013 hPa)
Melting Pt:–46 °C
Density:0,7822 g/cm³ (20 °C)
Flash Pt:6 °C
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
REACH:01-2119471307-38
:GHS08
:GHS07
:GHS02
CAS Number:75-05-8
Formula:H₃CCN
MDL Number:MFCD00001878