Acetonitrile ≥99.9%, B&J Brand™ for gas chromatography, for pesticide residue analysis, non-spectrophotometric grade, Burdick & Jackson™
BJAA016-4
:
- Pack type:Glass bottle
- Pk:16 L
- Seq:5254
- Electron Capture GC:10 ng/L max.
- GC Analysis:99.9 % min.
- Refractive Index (20ºC):1.3434 - 1.3446
- Residue:1 mg/L max.
- Water by Karl Fischer Titration:0.05 % max.
:
:41.05 g/mol
:13,00071
:82 °C (1,013 hPa)
:–46 °C
:0,7822 g/cm³ (20 °C)
:6 °C
:Ambient
:200-835-2
:1648
:3,II
:01-2119471307-38
:GHS08
:GHS07
:GHS02
:75-05-8
:H₃CCN
:MFCD00001878