Chlorobenzene ≥99.9%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™
Catalog # BJAA043-4
Supplier: BURDICK JACKSON
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:16 L
- Seq:5254
Specifications
Specification Test Results
- GC Analysis:99.9 % min.
- Refractive Index (20ºC):1.5242 - 1.5256
- UV Absorbance @ 300 nm:0.05 AU max.
- UV Absorbance @ 325 nm:0.04 AU max.
- UV Absorbance @ 350 nm:0.02 AU max.
- UV Absorbance @ 400 nm:0.005 AU max.
- UV Cutoff:287 nm max.
- Water by Karl Fischer Titration:0.03 % max.
Specification Test Results
About this item
:
:112.56 g/mol
Merck Index:13,02139
Boiling Pt:132 °C
Melting Pt:–46 °C
Density:1,1058 g/cm³ (20 °C)
Flash Pt:28 °C
Storage Temperature:Ambient
EINECS:203-628-5
UN:1134
ADR:3,III
REACH:01-2119432722-45
:GHS02
:GHS07
:GHS09
CAS Number:108-90-7
Formula:C₆H₅Cl
MDL Number:MFCD00000530