

4-Methyl-2-pentanone ≥99.0%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™
Catalog # BJAA253-4
Supplier: BURDICK JACKSON
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:16 L
- Seq:5254
Specifications
Specification Test Results
- Refractive Index (20ºC):1.3953 - 1.3965
- UV Absorbance @ 340 nm:0.5 AU max.
- UV Absorbance @ 350 nm:0.25 AU max.
- UV Absorbance @ 375 nm:0.05 AU max.
- UV Absorbance @ 400 nm:0.005 AU max.
- UV Cutoff:334 nm max.
- Water by Karl Fischer Titration:0.05 % max.
Specification Test Results
About this item
:
:100.16 g/mol
Merck Index:13,05227
Boiling Pt:116,5 °C
Melting Pt:–84 °C
Density:0,80 g/cm³ (20 °C)
Flash Pt:12,78 °C
Storage Temperature:Ambient
EINECS:203-550-1
UN:1245
ADR:3,II
:GHS07
:GHS02
CAS Number:108-10-1
Formula:(CH₃)₂CHCH₂COCH₃
MDL Number:MFCD00008938