
Methyl-2-chloro-4-fluorobenzoate ≥97%
Catalog # ALFAA18342.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:188.59
Boiling Pt:103…105 °C (15 mmHg)
:GHS07
Storage Temperature:Ambient
CAS Number:85953-29-3
Formula:C₈H₆ClFO₂
MDL Number:MFCD00173937