1-Propanol ≥99.5%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™
Catalog # BJAA322-4
Supplier: BURDICK JACKSON
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:16 L
- Seq:5254
Specifications
Specification Test Results
- GC Analysis:99.5 % min.
- Refractive Index (20ºC):1.3841 - 1.3857
- Residue:3 mg/L max.
- UV Absorbance @ 225 nm:0.5 AU max.
- UV Absorbance @ 250 nm:0.05 AU max.
- UV Absorbance @ 300 nm:0.005 AU max.
- UV Absorbance @ 400 nm:0.005 AU max.
- UV Cutoff:210 nm max.
- Water by Karl Fischer Titration:0.05 % max.
Specification Test Results
About this item
:
:60.1 g/mol
Merck Index:13,07934
Boiling Pt:97,2 °C
Melting Pt:-126,2 °C
Melting Pt:–126,2 °C
Density:0,8053 g/cm³ (20 °C)
Flash Pt:13,7 %(V)
EINECS:200-746-9
UN:1274
ADR:3,III
:GHS07
:GHS05
:GHS02
Storage Temperature:Ambient
CAS Number:71-23-8
Formula:H₃CCH₂CH₂OH
MDL Number:MFCD00002941