

1-Adamantanecarbonitrile ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:23074-42-2
:
Formula:C₁₁H₁₅N
MDL Number:MFCD00074731
:161.25 g/mol
Boiling Pt:275 °C (1013 hPa)
Melting Pt:193…196 °C
Density:1,13 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:245-413-9
UN:3439
ADR:6.1,III
:GHS06
:GHS07