3-Chloro-4-methoxybenzonitrile ≥97%
Catalog # ALFAL18509.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:167.59 g/mol
Boiling Pt:276 °C (1013 hPa)
Melting Pt:108…109 °C
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
:GHS06
:GHS07
CAS Number:102151-33-7
Formula:C₈H₆ClNO
MDL Number:MFCD03093073