Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Amantadine hydrochloride
Amantadine hydrochloride
Amantadine hydrochloride
EDQMA0363000
:
EDQM
:
Amantadine hydrochloride
EDQMA0363000
:
EDQM
:
Pack type:
Glass bottle for solids
Pk:
100 mg
Seq:
2440
:
:
187.71 g/mol
:
360 °C
:
211-560-2
:
665-66-7
:
C₁₀H₁₈ClN
:
MFCD00074723