1,2,4-Trichlorobenzene ≥98.0%, B&J Brand™ for GPC (gel permeation chromatography), for gas chromatography, for spectrophotometry, Burdick & Jackson™
Catalog # BJAA348-4
Supplier: BURDICK JACKSON
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:16 L
- Seq:5254
Specifications
Specification Test Results
- GC Analysis:98 % min.
- Refractive Index (20ºC):1.5711 - 1.5721
- UV Absorbance @ 310 nm:0.5 AU max.
- UV Absorbance @ 350 nm:0.05 AU max.
- UV Absorbance @ 375 nm:0.01 AU max.
- UV Absorbance @ 400 nm:0.005 AU max.
- UV Cutoff:308 nm max.
- Water by Karl Fischer Titration:0.01 % max.
Specification Test Results
About this item
:
:181.45 g/mol
Merck Index:13,09704
Boiling Pt:213,5 °C
Melting Pt:17 °C
Density:1,454 g/cm³ (20 °C)
Flash Pt:6,6 %(V) (150 °C)
Storage Temperature:Ambient
EINECS:204-428-0
UN:2321
ADR:6.1,III
:GHS07
:GHS09
CAS Number:120-82-1
Formula:C₆H₃Cl₃
MDL Number:MFCD00000547