Tubing connectors, valved, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
PP, beige.
- Fluid flow stops when fitting halves are separated
- Designed for one-handed quick disconnect with flexible tubing
- Provides a leakproof† connection between two tubes, but may require clamps
Maintain the integrity of transfer liquids. The barbed fitting on each end and internal valve in the middle, which seals when the fittings are removed, are ideal for use in developing in-house fluid transfer systems.
Specifications
- Catalog No:
- NALG6177-0250
- NALG6177-0375
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement