


Acetonitrile ≥99.9%, BioSyn™, meets UV Specifications for biosynthesis, for applications requiring low water content, Burdick & Jackson™
Supplier: BURDICK JACKSON
Certificates
About this item
CAS Number:75-05-8
:
Formula:H₃CCN
MDL Number:MFCD00001878
:41.05 g/mol
Merck Index:13,00071
Boiling Pt:82 °C (1,013 hPa)
Melting Pt:–46°C
Density:0,7857 g/cm³ (20 °C)
Flash Pt:6 °C
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
REACH:01-2119471307-38
:GHS07
:GHS08
:GHS02
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
- GC Analysis:99.9 % min.
- Refractive Index (20ºC):1.3434 - 1.3446
- Residue:1 mg/L max.
- UV Absorbance @ 200 nm:0.05 AU max.
- UV Absorbance @ 210 nm:0.04 AU max.
- UV Absorbance @ 225 nm:0.01 AU max.
- UV Cutoff:190 nm max.
- Water by Karl Fischer Titration:20 ppm max.