

1-Iodobutane ≥99% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:542-69-8
:
Formula:C₄H₉I
MDL Number:MFCD00001098
:184.02 g/mol
Merck Index:14,1573
Boiling Pt:129…131 °C
Melting Pt:–103 °C
Density:1,616
Flash Pt:33 °C (91 °F)
UN:1993
ADR:3,III
:Stabilised with copper
EINECS:208-824-4
:GHS02
:GHS07
Storage Temperature:Ambient