Centrifuge tubes, cylindrical, round bottom
Supplier: Brand
Certificates
About this item
PP, translucent.
- Translucent, rimmed, very good chemical resistance
- Autoclavable at 121 °C (2 bar), according DIN EN 285
- Fitting stoppers must be ordered separately