Sodium hydroxide 0.5 mol/l (0.5 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur. volumetric solution
: Caustic soda
:1310-73-2
:
:NaOH
:MFCD00003548
:40 g/mol
:>105 °C (1013 hPa)
:1,01 g/cm³ (20 °C)
:Ambient
:215-185-5
:1824
:8,III
:GHS05
- Environmentally Preferable:
- NIST traceable:Confirmed
- Titer (20°C; real value 0.2 % accuracy):0.499 - 0.501 mol/l