Order Entry
Singapore
ContactUsLinkComponent
Orthophosphoric acid ≥85%, ULTREX® II, Ultrapure, J.T.Baker®
Corrosive
Orthophosphoric acid ≥85%, ULTREX® II, Ultrapure, J.T.Baker®
Catalog # BAKR6908-04
Supplier:  Avantor
CAS Number:  
undefined
Corrosive
Orthophosphoric acid ≥85%, ULTREX® II, Ultrapure, J.T.Baker®
Catalog # BAKR6908-04
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 g
  • Seq:
    3368

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    Passes Test
  • Appearance:
    Passes Test
  • Assay (H₃PO₄)(w/w), For Information Only:
    For Information Only
  • Density (g/mL) at 25°C, For Information Only:
    For Information Only
  • Trace Impurities - Arsenic (As), For Information Only:
    For Information Only
  • Trace Impurities - Boron (B), For Information Only:
    For Information Only
  • Nitrate (NO₃), For Information Only:
    For Information Only
  • Sulfate (SO₄), For Information Only:
    For Information Only
  • Trace Impurities - Aluminum (Al), For Information Only:
    For Information Only
  • Trace Impurities - Antimony (Sb), For Information Only:
    For Information Only
  • Trace Impurities - Barium (Ba), For Information Only:
    For Information Only
  • Trace Impurities - Beryllium (Be), For Information Only:
    For Information Only
  • Trace Impurities - Bismuth (Bi), For Information Only:
    For Information Only
  • Trace Impurities - Cadmium (Cd), For Information Only:
    For Information Only
  • Trace Impurities - Calcium (Ca), For Information Only:
    For Information Only
  • Trace Impurities - Cerium (Ce), For Information Only:
    For Information Only
  • Trace Impurities - Cesium (Cs), For Information Only:
    For Information Only
  • Trace Impurities - Chromium (Cr), For Information Only:
    For Information Only
  • Trace Impurities - Cobalt (Co), For Information Only:
    For Information Only
  • Trace Impurities - Copper (Cu), For Information Only:
    For Information Only
  • Trace Impurities - Dysprosium (Dy), For Information Only:
    For Information Only
  • Trace Impurities - Erbium (Er), For Information Only:
    For Information Only
  • Trace Impurities - Europium (Eu), For Information Only:
    For Information Only
  • Trace Impurities - Gadolinium (Gd), For Information Only:
    For Information Only
  • Trace Impurities - Gallium (Ga), For Information Only:
    For Information Only
  • Trace Impurities - Germanium (Ge), For Information Only:
    For Information Only
  • Trace Impurities - Gold (Au), For Information Only:
    For Information Only
  • Trace Impurities - Holmium (Ho), For Information Only:
    For Information Only
  • Trace Impurities - Indium (In), For Information Only:
    For Information Only
  • Trace Impurities - Iridium (Ir), For Information Only:
    For Information Only
  • Trace Impurities - Iron (Fe), For Information Only:
    For Information Only
  • Trace Impurities - Lanthanum (La), For Information Only:
    For Information Only
  • Trace Impurities - Lead (Pb), For Information Only:
    For Information Only
  • Trace Impurities - Lithium (Li), For Information Only:
    For Information Only
  • Trace Impurities - Lutetium (Lu), For Information Only:
    For Information Only
  • Trace Impurities - Magnesium (Mg), For Information Only:
    For Information Only
  • Trace Impurities - Manganese (Mn), For Information Only:
    For Information Only
  • Trace Impurities - Mercury (Hg), For Information Only:
    For Information Only
  • Trace Impurities - Molybdenum (Mo), For Information Only:
    For Information Only
  • Trace Impurities - Neodymium (Nd), For Information Only:
    For Information Only
  • Trace Impurities - Nickel (Ni), For Information Only:
    For Information Only
  • Trace Impurities - Niobium (Nb), For Information Only:
    For Information Only
  • Trace Impurities - Palladium (Pd), For Information Only:
    For Information Only
  • Trace Impurities - Platinum (Pt), For Information Only:
    For Information Only
  • Trace Impurities - Potassium (K), For Information Only:
    For Information Only
  • Trace Impurities - Praseodymium (Pr), For Information Only:
    For Information Only
  • Trace Impurities - Rhodium (Rh), For Information Only:
    For Information Only
  • Trace Impurities - Rubidium (Rb), For Information Only:
    For Information Only
  • Trace Impurities - Ruthenium (Ru), For Information Only:
    For Information Only
  • Trace Impurities - Samarium (Sm), For Information Only:
    For Information Only
  • Trace Impurities - Silver (Ag), For Information Only:
    For Information Only
  • Trace Impurities - Sodium (Na), For Information Only:
    For Information Only
  • Trace Impurities - Strontium (Sr), For Information Only:
    For Information Only
  • Trace Impurities - Tantalum (Ta), For Information Only:
    For Information Only
  • Trace Impurities - Terbium (Tb), For Information Only:
    For Information Only
  • Trace Impurities - Thallium (Tl), For Information Only:
    For Information Only
  • Trace Impurities - Thorium (Th), For Information Only:
    For Information Only
  • Trace Impurities - Thulium (Tm), For Information Only:
    For Information Only
  • Trace Impurities - Tin (Sn), For Information Only:
    For Information Only
  • Trace Impurities - Titanium (Ti), For Information Only:
    For Information Only
  • Trace Impurities - Tungsten (W), For Information Only:
    For Information Only
  • Trace Impurities - Uranium (U), For Information Only:
    For Information Only
  • Trace Impurities - Vanadium (V), For Information Only:
    For Information Only
  • Trace Impurities - Ytterbium (Yb), For Information Only:
    For Information Only
  • Trace Impurities - Yttrium (Y), For Information Only:
    For Information Only
  • Trace Impurities - Zinc (Zn), For Information Only:
    For Information Only

Specification Test Results

About this item

 :
 :98 g/mol
Melting Pt:28 °C
Density:1,71…1.87 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-633-2
UN:1805
ADR:8,III
 :GHS05
CAS Number:7664-38-2
Formula:H₃PO₄
MDL Number:MFCD00011340