Tubings, polyurethane, Nalgene®
Polyurethane, translucent.
- Contains no plasticizers and only low levels of extractables for high-purity applications
- Superior chemical resistance to fuels, oils, and some solvents (not recommended for aqueous solutions)
- Resistant to atmospheric ozone, aliphatic hydrocarbons and petroleum products for longer life and versitility
- This transparent, non-autoclavable tubing is marked every 12" for cutting and can be gas sterilized
- Ideal for peristaltic pump applications
Polyurethane tubing, which exhibits excellent abrasion and flex-fatigue resistance, as well as low-temperature resilience.
- Catalog No:
- NALG8030-0060
- NALG8030-0120
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement