Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Hydrochloric acid
Hydrochloric acid 6 mol/l (6 N), Reagent Grade
Hydrochloric acid 6 mol/l (6 N), Reagent Grade
:
VWR Chemicals
Hydrochloric acid 6 mol/l (6 N), Reagent Grade
:
VWR Chemicals
:
7647-01-0
:
:
HCl
:
MFCD00011324
:
1,06 g/cm³ (20 °C)
:
Ambient
:
231-595-7
:
1789
:
8,II
:
36.46
:
GHS07
:
GHS05
Concentration:
5.4 – 6.6 moles/l
SDS_EN_SG_VWRCE484.pdf
sds_VWRCE484