Hydrochloric acid ≥30%, TraceSELECT™ for trace analysis, Fluka™
Supplier: Honeywell Chemicals
Hydrochloric acid ≥30%, TraceSELECT™ for trace analysis, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
TraceSELECT™ Ultra and TraceSELECT
- Ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- Sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- The acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (µg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions
Synonyms: Hydrochloric acid solution, Muriatic acid
- CAS Number:7647-01-0
- Formula:HCl
- MDL Number:MFCD00011324
- Molecular Weight:36.46
- Boiling Pt:42 °C (1,013 hPa)
- Melting Pt:–35 °C
- Density:1,190 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-595-7
- UN:1789
- ADR:8,II
4 Options Available Below
Product Details & Documents
TraceSELECT™ Ultra and TraceSELECT
- Ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- Sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- The acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (µg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions
Synonyms: Hydrochloric acid solution, Muriatic acid
- CAS Number:7647-01-0
- Formula:HCl
- MDL Number:MFCD00011324
- Molecular Weight:36.46
- Boiling Pt:42 °C (1,013 hPa)
- Melting Pt:–35 °C
- Density:1,190 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-595-7
- UN:1789
- ADR:8,II
Specifications for Product Family
Specification Test Results
- Assay:30.0 - 32.0 %
- Density (D 20/4):1.140 - 1.160
- Silver (Ag):Max. 0.5 ppb
- Aluminium (Al):Max. 5 ppb
- Arsenic (As):Max. 1 ppb
- Gold (Au):Max. 1 ppb
- Barium (Ba):Max. 0.5 ppb
- Beryllium (Be):Max. 0.5 ppb
- Bismuth (Bi):Max. 0.5 ppb
- Calcium (Ca):Max. 10 ppb
- Cadmium (Cd):Max. 0.5 ppb
- Cobalt (Co):Max. 0.5 ppb
- Chromium (Cr):Max. 1 ppb
- Cesium (Cs):Max. 0.5 ppb
- Copper (Cu):Max. 1 ppb
- Iron (Fe):Max. 5 ppb
- Gallium (Ga):Max. 0.5 ppb
- Mercury (Hg):Max. 0.5 ppb
- Indium (In):Max. 0.5 ppb
- Iridium (Ir):Max. 0.5 ppb
- Potassium (K):Max. 10 ppb
- Lithium (Li):Max. 0.5 ppb
- Magnesium (Mg):Max. 1 ppb
- Manganese (Mn):Max. 0.5 ppb
- Molybdenum (Mo):Max. 0.5 ppb
- Sodium (Na):Max. 10 ppb
- Nickel (Ni):Max. 1 ppb
- Lead (Pb):Max. 0.5 ppb
- Palladium (Pd):Max. 1 ppb
- Platinum (Pt):Max. 0.5 ppb
- Rubidium (Rb):Max. 0.5 ppb
- Rhodium (Rh):Max. 0.5 ppb
- Ruthenium (Ru):Max. 0.5 ppb
- Antimony (Sb):Max. 0.5 ppb
- Selenium (Se):Max. 0.001 ppm
- Tin (Sn):Max. 1 ppb
- Strontium (Sr):Max. 0.5 ppb
- Titanium (Ti):Max. 1 ppb
- Thallium (Tl):Max. 0.5 ppb
- Uran (U):Max. 0.5 ppb
- Vanadium (V):Max. 1 ppb
- Zinc (Zn):Max. 1 ppb
- Zirconium (Zr):Max. 0.5 ppb
- Bromide (Br):Max. 10000 ppb
- Free chlorine (Cl):Max. 0.1 ppm
- Nitrate (NO₃):Max. 1000 ppb
- Phosphate (PO₄):Max. 10 ppb
- Sulfite (SO₃):Max. 1 ppm
- Sulfate (SO₄):Max. 100 ppb



