Hydrochloric acid 24,5 - 26,0%, Puriss., Fluka™
: Hydrochloric acid solution, Muriatic acid
:7647-01-0
:
:HCl
:MFCD00011324
:1.11 g/cm3
:36.46
:Ambient
:231-595-7
:1789
:8 II
:GHS05
:GHS07
- Environmentally Preferable:
- Ignition Residue:≤0.002% (as SO₄)
- Assay:24.5 - 26.0 %
- Residue on ignition (SO₄):Max. 0.002 %
- Arsenic (As):Max. 0.00005 %
- Iron (Fe):Max. 0.0001 %
- Heavy metals (as Pb):Max. 0.0001 %
- Free chlorine (Cl):Max. 0.0001 %
- Sulfate (SO₄):Max. 0.0002 %
- Sulfite (SO₃):Max. 0.0005 %
- Vapor Pressure:7.93 psi ( 37.7 °C)