Copper ≥99.999% (metals basis), wire, Puratronic®, Ø 2.0 mm (0.08 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-50-8
:
Formula:Cu
MDL Number:MFCD00010965
:63.55 g/mol
Merck Index:13,02545
Boiling Pt:2595 °C (1013 hPa)
Melting Pt:1083 °C
Density:8,94 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-159-6