Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Copper
Copper ≥99.9985% (metals basis), wire, Puratronic®, Ø 0.25 mm (0.01 in)
Copper ≥99.9985% (metals basis), wire, Puratronic®, Ø 0.25 mm (0.01 in)
:
Thermo Fisher Scientific
Copper ≥99.9985% (metals basis), wire, Puratronic®, Ø 0.25 mm (0.01 in)
:
Thermo Fisher Scientific
:
7440-50-8
:
:
Cu
:
MFCD00010965
:
63.55 g/mol
:
13,02545
:
2595 °C (1013 hPa)
:
1083 °C
:
8,94 g/cm³ (20 °C)
:
Ambient
:
231-159-6
msds_ALFA010972