Order Entry
Singapore
Orders LinkContactUsLinkComponent
Capsaicin

Capsaicin

Glass bottle for solids, 50 mg
Catalog # EDQMY0000671
Supplier:   EDQM
Capsaicin
Catalog # EDQMY0000671
Supplier:  EDQM
CAS Number:  404-86-4
Capsaicin

Glass bottle for solids, 50 mg

CAS Number:  404-86-4
Price
$1,940.00
Log in for availability
1 * 50 mg
Product Details & Documents
Synonyms:  8-Methyl-N-vanillyl-6-none-amid
  • CAS Number:404-86-4
  • Formula:C₁₈H₂₇NO₃
  • MDL Number:MFCD00017259
  • Molecular Weight:305.42 g/mol
  • Merck Index:12,01811
  • Boiling Pt:210…220 °C (0,01 mmHg)
  • Melting Pt:66,1 °C
  • EINECS:206-969-8
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 mg

Specifications