

Diethyl ether ≥99.9% contains no preservatives, B&J Brand™ for HPLC, for pesticide residue analysis, for spectrophotometry, Burdick & Jackson™
BJAA107-4
:
- Pack type:Glass bottle
- Pk:16 L
- Seq:5254
- Electron Capture GC:10 ng/L max.
- Water by Karl Fischer Titration:0.03 % max.
- GC Analysis:99.9 % min.
- Peroxides (as H₂O₂) at time of packaging:1 ppm max.
- Refractive Index (20ºC):1.3516 - 1.3526
- Residue:1 mg/L max.
- UV Absorbance @ 250 nm:0.08 AU max.
- UV Absorbance @ 275 nm:0.01 AU max.
- UV Absorbance @ 300 nm:0.005 AU max.
- UV Cutoff:215 nm max.
:
:74.12 g/mol
:12,03852
:35 °C
:–117,4 °C
:0,713 g/cm³ (20 °C)
:–45 °C
:Ambient
:200-467-2
:1155
:3,I
:01-2119535785-29
:GHS07
:GHS02
:60-29-7
:(CH₃CH₂)₂O
:MFCD00011646