Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Giemsa's stain
Giemsa's stain stock solution
Giemsa's stain stock solution
Giemsa's stain stock solution
RICCR3250000-120A
:
Ricca Chemical
:
Giemsa's stain stock solution
RICCR3250000-120A
:
Ricca Chemical
:
Pack type:
Glass bottle
Pk:
120 mL
Seq:
2470
:
:
0,99 g/cm³ (20 °C)
:
Ambient
:
257-438-2
:
1993
:
3,II
:
GHS02
:
GHS06
:
GHS08
:
GHS07
:
51811-82-6
:
MFCD00081642