Order Entry
Singapore
Orders LinkContactUsLinkComponent
Programmable time switch
Programmable time switch
Catalog # IBSAETU17
Supplier:   IBS Stock Items
Programmable time switch
Catalog # IBSAETU17
Supplier:  IBS Stock Items
Programmable time switch
Programmable time switch
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Large LCD shows time, On/Off status and date. Random switching facility and programme override.
  • Simple two button programming
  • Six on/off operations per 24 hours for 7 days
  • 24 hours / 7 days (weekday and weekend) programmes
Plugs into 13 A power socket, maximum 3000 W loading.