VWR® DRY-Line®, Ovens with Natural Convection
Supplier: VWR Collection
Certificates
About this item
Drying ovens with natural convection used for thermal processes, heated storage and for convection drying applications with temperatures up to 220 °C. Exhaust duct at the back of the unit with manually adjustable slide.
- Microprocessor controller
- Integrated timer can be set for continuous mode or timer operation: 0 to 999 min or 0 to 99,9 h
- Over-temperature protection with visual alarm (Class 3.1 safety device)
- Temperature setting in increments of 1 °C
Delivery information: Supplied with two chrome plated shelves.