Order Entry
Singapore
Orders LinkContactUsLinkComponent
Copan Transystem® Traditional Swab Collection and Transport System for Aerobic and Anaerobic Bacteria
Copan Transystem® Traditional Swab Collection and Transport System for Aerobic and Anaerobic Bacteria
Supplier:   Copan
Copan Transystem® Traditional Swab Collection and Transport System for Aerobic and Anaerobic Bacteria
Supplier:  Copan
Certificates
Copan Transystem® Traditional Swab Collection and Transport System for Aerobic and Anaerobic Bacteria
Copan Transystem® Traditional Swab Collection and Transport System for Aerobic and Anaerobic Bacteria
13 Options Available Below
Product Details & Documents
Our Transystem™ family comprises different media for the efficient and safe transport of many bacterial strains. Choose between liquid or solid Amies and Stuart medium for aerobic culture, rapid antigen, and molecular testing; opt for gel Cary-Blair medium - with or without charcoal - for aerobic and anaerobic cultures. Bacterial transport basics. A simple, low bioburden, device for every investigation. Just uncap, collect and recap the tube.
  • Swab variety: Various swab sizes to reach different collection sites
    Media assortment: Choice of liquid or solid Amies and Stuart medium for aerobic culture, rapid antigen, and molecular testing. Choice of Amies, Stuart, or Cary-Blair Gel medium (with or without charcoal), commonly used for aerobic and anaerobic cultures
    Venturi hourglass tube: The Venturi unique 'hourglass' design eliminates air pockets - harmful to anaerobe bacteria - preventing dehydration and breakdown of the gel column
    Proprietary state of the art outer packaging: The VI-PAK metallic foil and plastic film barrier block oxygen entry, preventing unwanted oxidation of the transport medium
This product line groups a variety of product codes, each optimised to address specific needs.
Application: Culture - Clinical bacteriology
Product Family Options
Product Information
  • Cap colour
    Availability
    Price
  • COPA111C.USE
    Transystem™ - plastic applicator, rayon wrapped regular tip, stuart without charcoal, choked tube, 139 mm, 5 ml, 50×8, individually wrapped
    Transystem™ - plastic applicator, rayon wrapped regular tip, stuart without charcoal, choked tube, 139 mm, 5 ml, 50×8, individually wrapped
    Blue
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
  • COPA117C.USE
    Transystem™ - plastic applicator, rayon wrapped regular tip, stuart with charcoal, choked tube, 139 mm, 50×8, individually wrapped
    Transystem™ - plastic applicator, rayon wrapped regular tip, stuart with charcoal, choked tube, 139 mm, 50×8, individually wrapped
    Black
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
  • COPA132C.USE
    Transystem™ - plastic applicator, rayon wrapped regular tip, cary blair medium, choked tube, 139 mm, 50×8, individually wrapped
    Transystem™ - plastic applicator, rayon wrapped regular tip, cary blair medium, choked tube, 139 mm, 50×8, individually wrapped
    Red
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
  • COPA134C.USE
    Transystem™ - dual swab plastic applicators, rayon wrapped regular tip, amies without charcoal, choked tube, 139 mm, 50×10, individually wrapped
    Transystem™ - dual swab plastic applicators, rayon wrapped regular tip, amies without charcoal, choked tube, 139 mm, 50×10, individually wrapped
    Blue
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
  • COPA136C.USE
    Transystem™ - dual swab plastic applicators, rayon wrapped regular tip, amies with charcoal, choked tube, 139 mm, 50×10, individually wrapped
    Transystem™ - dual swab plastic applicators, rayon wrapped regular tip, amies with charcoal, choked tube, 139 mm, 50×10, individually wrapped
    White
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
  • COPA113C.USE
    Transystem™ - aluminium applicator, rayon wrapped minitip, stuart without charcoal, choked tube, 139 mm, 50×8, individually wrapped
    Transystem™ - aluminium applicator, rayon wrapped minitip, stuart without charcoal, choked tube, 139 mm, 50×8, individually wrapped
    Orange
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
  • COPA125C.USE
    Transystem™ - soft aluminium applicator, rayon wrapped minitip, amies with charcoal, choked tube, 139 mm, 50×10, individually wrapped
    Transystem™ - soft aluminium applicator, rayon wrapped minitip, amies with charcoal, choked tube, 139 mm, 50×10, individually wrapped
    Green
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
  • COPA143C.USE
    Transystem™ - aluminium applicator, rayon wrapped minitip, stuart liquid medium without charcoal, choked tube, 139 mm, 50×8, individually wrapped
    Transystem™ - aluminium applicator, rayon wrapped minitip, stuart liquid medium without charcoal, choked tube, 139 mm, 50×8, individually wrapped
    Orange
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
  • COPA124C.USE
    Transystem™ - soft aluminium applicator, rayon wrapped minitip, amies without charcoal, choked tube, 139 mm, 50×10, individually wrapped
    Transystem™ - soft aluminium applicator, rayon wrapped minitip, amies without charcoal, choked tube, 139 mm, 50×10, individually wrapped
    Green
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
  • COPA144C
    Transystem™ - chlamydia swab plastic applicator, polyester wrapped regular tip, liquid medium, choked tube, 139 mm, 25×10, individually wrapped
    Transystem™ - chlamydia swab plastic applicator, polyester wrapped regular tip, liquid medium, choked tube, 139 mm, 25×10, individually wrapped
    Yellow
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items
  • COPA147C
    Transystem™ - viro swab plastic applicator, polyester wrapped regular tip, liquid medium, choked tube, 139 mm, 25×10, individually wrapped
    Transystem™ - viro swab plastic applicator, polyester wrapped regular tip, liquid medium, choked tube, 139 mm, 25×10, individually wrapped
    Pink
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items
  • COPA148C
    Transystem™ - viro swab aluminium applicator, polyester wrapped regular tip, liquid medium, choked tube, 139 mm, 25×10, individually wrapped
    Transystem™ - viro swab aluminium applicator, polyester wrapped regular tip, liquid medium, choked tube, 139 mm, 25×10, individually wrapped
    Pink
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items
  • COPA145C
    Transystem™ - chlamydia swab aluminium applicator, polyester wrapped regular tip, liquid medium, choked tube, 139 mm, 25×10, individually wrapped
    Transystem™ - chlamydia swab aluminium applicator, polyester wrapped regular tip, liquid medium, choked tube, 139 mm, 25×10, individually wrapped
    Yellow
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items