Order Entry
Singapore
ContactUsLinkComponent
Transport swabs
Transport swabs
Catalog # SLIN190C.USE_
Supplier:  Sterilin
undefined
Transport swabs
Catalog # SLIN190C.USE_
Supplier:  Sterilin
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Transport swabs
  • Seq:
    06
  • Shaft/head:
    Twisted wire/rayon
  • Transport medium:
    Amies gel
  • Cap colour:
    Blue
  • Pk:
    500
Specifications
About this item
Designed for sample collection and transport of bacteria, these swabs contain media for the maintenance of bacteria during transport to the laboratory.
  • Viability of bacteria for 24 to 48 hours
  • Pouch seal turning white on opening as a visible sign of tamper evidence
  • Printed with lot number and expiry date and tamper evident sleeve ensuring traceability and sterility
Tested for performance using a wide range of aerobic and anaerobic organisms ensuring adequate recovery together with sterility and other quality assurance tests.
Certifications: Class lla in accordance with Medical Device Directive 93/42/EEC (for transient invasive use).
Packaging: Unique nitrogen flushed aluminium inner bag with laminated film pouch to prevent fromoxidation and dehydration of media.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904