Order Entry
Singapore
ContactUsLinkComponent
Pipette controller, Bibbette, Azlon®
Pipette controller, Bibbette, Azlon®
Catalog # AZLOPFL001N
Supplier:  SciLabware
undefined
Pipette controller, Bibbette, Azlon®
Catalog # AZLOPFL001N
Supplier:  SciLabware
Specifications
  • Description:
    Pipette controller Bibbette, white
  • Seq:
    02
  • Pk:
    1
Specifications
About this item
Pipette controller for use with pipettes from 0,1 up to 100 ml (accommodated by silicone adapter).
  • Portable, lightweight
  • Single handed operation
  • Comfortable control of the filling, volume adjustment and dispensing of liquids through thumb-operated controls
  • Fine level adjustment and blow through for blow-out pipettes