Knitted gloves, Tiger Paw® 76-301
: Ansell
Made from knitted polyester coated with cotton, Tiger Paw® offers many of the features and benefits of Tri Grip™ at a more economical price. This heavier construction ensures a more robust glove for heavier applications.
- Seamless construction avoids irritation to fingers caused by seams
- Elastic wrist ensures glove remains in constant fixed position
- Knitted construction allows the air to circulate thus reducing perspiration
- PVC polka dots provide a secure grip without loss of dexterity
- Machine washable at 40 °C
: EN 388:2016 (214XB), Cat. II
- Catalog No:
- ANSE76-301/10
- ANSE76-301/7
- Model:
- 76-301
- 76-301
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Liner material:
- Polyester/cotton
- Polyester/cotton
- Finish:
- Dot coating on palm
- Dot coating on palm
- Gauge:
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (214XB)
- EN 388:2016 (214XB)