Capillary columns, BP1
Supplier: Trajan Scientific and Medical
Certificates
About this item
These 100% dimethyl polysiloxane columns are suitable for the analysis of hydrocarbons, aromatics, pesticides, phenol, herbicides and amines.
- Classic dimethyl polysiloxane technology with high temperature cross-linking
- Non polar phase suitable for all routine analyses
- Low bleed
Suitable replacement for: DB-1, DB-Petro, HP-1, HP-1MS, Rtx-1, Ultra-1, SPB-1, SPB-1 Sulfur, Petrocol DH, CP-Sil 5CB, VB-1, ZB-1, VF-1ms.
Max. temperature: 360 °C.
100% dimethyl polysiloxane
An non-polar phase for PONA analysis.