Chemical resistant overalls, Tychem® 6000 F, models CHA5 / CHA6
: DuPont
Tychem® 6000 F overalls are made from a lightweight and durable fabric (<500 g per garment) consisting of a proprietary barrier film laminated to a heavy duty Tyvek® substrate. They offer excellent chemical permeation protection to an extensive range of chemicals, helping to protect personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents.
- With hood; elasticated face, wrists, waist and ankles for an optimal fit
- Self-adhesive double zip flap closure system for higher protection
- Self-adhesive chin flap for tight seal of suit to face-masks
- Thumb loops prevent sleeves from riding up
- Stitched and over-taped seams with barrier tape for protection and strength
- Anti-static treatment (EN 1149-5) on inside
- Available in grey or in orange for high visibility
- CHA6: With integrated socks to be worn inside shoes or boots and additional knee-length boot flaps
Tychem® 6000 F is used for a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications.
Colour: Grey or orange.
Weight: 120 g/m².
: Type 3-B/4-B/5-B/6-B, EN 1149-5, EN 1073-2, EN 14126, Cat. III.; (EU) 2016/425
: 25 per box, individually packed.
- Catalog No:
- DUPOTFCHA5TGYSM002
- DUPOCHA5TGY16/XXXL
- DUPOTFCHA5TGYLG002
- DUPOCHA5TOR00/XL
- DUPOTFCHA5TGYMD002
- DUPOCHA5TGY16/XL
- DUPOCHA5TGY16/XXL
- DUPOTFCHA5TGYXL002
- DUPOTYFCHA5TOR00/M
- DUPOTFCHA5TGY2X002
- DUPOTYFCHA5TGY16/S
- DUPOTYFCHA5TOR00/L
- DUPOTYFCHA5TGY16/L
- DUPOTYFCHA5TGY16/M
- DUPOCHA5TOR00/XXL
- DUPOTFCHA5TGY3X002
- Model:
- 6000 F
- 6000 F
- 6000 F
- 6000 F
- 6000 F
- 6000 F
- 6000 F
- 6000 F
- 6000 F
- 6000 F
- 6000 F
- 6000 F
- 6000 F
- 6000 F
- 6000 F
- 6000 F
- Anti-static:
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
- Garment application:
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Weight:
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²
- 120 g/m²