Overalls, ProShield® 30, model CHF5a
Catalog # DUPOS305SWH00/XXXL
Supplier: DuPont
Specifications
- Clothing Size:3XL
- Anti-static:+
- Garment application:Chemical protection,Medical,Laboratory,Clinical
- Colour:White
- Seq:06
- Model:30
- Pk:100
- Weight:60 g
Specifications
About this item
These garments comprise a microporous film on a PP non woven fabric and combine good particle protection with high liquid repellency. They help provide a barrier against non hazardous dry particles down to one micron in size, are repellent to liquids, but offer only limited chemical permeation barrier to some commonly used low-concentration water-based chemicals. This material offers limited comfort: it is permeable to moisture vapour only and it is not air permeable.
- 3-piece hood and 3-piece gusset for optimal fit
- Nylon zip with flap
- Elasticated face, wrists, waist and ankles
- Anti-static treatment (EN 1149-5) on inside
- Stitched internal seams
ProShield® 30 overalls are an ideal choice for applications that are less demanding in terms of barrier, durability and comfort, for example general maintenance, hospitals and other industries.
Colour: White
Weight: 60 g/m²
Certifications: Type 5/6, EN 1149-5 (only inside), EN 1073-2; Cat. III