Order Entry
Singapore
ContactUsLinkComponent
Buckets, Specimen Collection with Lids
Buckets, Specimen Collection with Lids
  RICRS6106
undefined
Buckets, Specimen Collection with Lids
  RICRS6106

 

  • Capacity:
    16 L
  • Colour:
    White
  • Height:
    272 mm
  • Seq:
    06
  • Pk:
    5
  • Ø int.:
    323 mm

 

 

HDPE, white.

  • Round shape allows economical use of space, stackable
  • Handle grip for easy lifting and carrying (216-4527 and 216-4528 without handle)
  • Easily removable seal-tight lids