Order Entry
Singapore
Orders LinkContactUsLinkComponent
Brushes for test tubes
Brushes for test tubes
Supplier:   PINNACLE BRUSH
Brushes for test tubes
Supplier:  PINNACLE BRUSH
Certificates
Brushes for test tubes
Brushes for test tubes
2 Options Available Below
Product Details & Documents
Brushes with nylon bristles and cotton yarn tip, on galvanized wire, particularly suitable for small tubes.
Product Family Options
Product Information
  • Bristle material
    Brush head length
    Brush head Ø
    Handle material
    Total length
    Availability
    Price
  • Nylon
    75 mm
    17 mm
    Wire
    240 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items
  • Nylon
    75 mm
    24 mm
    Wire
    240 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items