Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Nitroterephthalic acid
Nitroterephthalic acid ≥99%
Nitroterephthalic acid ≥99%
Irritant
Nitroterephthalic acid ≥99%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
Nitroterephthalic acid ≥99%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
610-29-7
:
Formula:
C₈H₅NO₆
MDL Number:
MFCD00007141
Melting Pt:
270…272 °C
:
211.13 g/mol
EINECS:
210-217-4
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAB25043
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation