


Nitric acid 20% (v/v) in aqueous solution (1 + 4)
Catalog # RICCR5326000-2.5D
Supplier: Ricca Chemical
New Product
CAS Number:
Specifications
- Pack type:Safety-coated glass
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Appearance:Colorless to light yellow liquid
- Assay (vs. Sodium Hydroxide/Phenolphthalein):19.9-20.1 % (v/v)
Specification Test Results
About this item
:
:63.01
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8,II
:GHS05
:GHS08
:GHS06
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349