Order Entry
Singapore
ContactUsLinkComponent
Sodium hydroxide 20% (w/w) in aqueous solution Reag. Ph. Eur. 1081401
Corrosive
Sodium hydroxide 20% (w/w) in aqueous solution Reag. Ph. Eur. 1081401
Catalog # VWRC87933.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Sodium hydroxide 20% (w/w) in aqueous solution Reag. Ph. Eur. 1081401
Catalog # VWRC87933.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :40 g/mol
Boiling Pt:119…135 °C (1013 hPa)
Melting Pt:9-10 °C
Density:∼1,22 g/ml
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548