

Dichloromethane ≥99.9% stabilised, BioSyn™ for biosynthesis, for applications requiring low water content, Burdick & Jackson™
BJAABB301-4
:
- Pack type:Glass bottle
- Pk:16 L
- Seq:5254
- Acidity (as HCl):1 mg/L max.
- Chloride:10 mg/L max.
- GC Analysis:99.9 % min.
- Refractive Index (20ºC):1.4236 - 1.4246
- Residue:1 mg/L max.
- UV Absorbance @ 240 nm:0.1 AU max.
- UV Absorbance @ 250 nm:0.01 AU max.
- UV Cutoff:233 nm max.
- Water by Karl Fischer Titration:20 ppm max.
:
:84.93 g/mol
:12,06140
:40 °C
:–95 °C
:1,33 g/cm³
: Does not flash
:200-838-9
:1593
:6.1,III
:Stabilised with cyclohexene
:GHS07
:GHS08
:Ambient
:75-09-2
:CH₂Cl₂
:MFCD00000881