


Phenol ≥99.0% saturated with buffer for biotechnology pH 4.5
VWRC0981-400ML
:
- Pack type:Glass bottle
- Pk:400 mL
- Seq:2665
- O.D.@ 330 nm:≤0.2
- O.D.@ 405 nm:≤0.1
- O.D.@ 510 nm:≤0.1
- pH (Buffer) @25 °C:4.1 – 4.5
- Purity:≥99 %
:
:94.11 g/mol
:182 °C
:Ambient
:203-632-7
:2821
:6.1,II
:GHS06
:GHS05
:GHS08
:108-95-2
:C₆H₅OH
:MFCD00002143