Order Entry
Singapore
ContactUsLinkComponent
4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP), Reagent Grade
4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP), Reagent Grade
Catalog # VWRC0364-25G
Supplier:  VWR Chemicals
CAS Number:  
undefined
4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP), Reagent Grade
Catalog # VWRC0364-25G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Em (312 nm, 0.15M AMP Buffer, pH 10.25):
    >9800
  • Inorganic Phosphate (% by Weight):
    <1.0%
  • p-Nitrophenol:
    <0.1%

Specification Test Results

About this item

CAS Number:7647-14-5
 :4264-83-9
Formula:C₆H₄NNa₂O₆P·6H₂O
MDL Number:MFCD00066288
Melting Pt:300 °C
 :371.144 g/mol
Storage Temperature:Freezer
EINECS:224-246-5