Micro-Porous Boiling Chips
WALU501A_B
New Product
- Description:Micro-porous boiling chips
- Pack type:227 g
- Pk:1
Treated, micro-porous, chemically inert carbon chips eliminate 'bumping' in either aqueous or non-aqueous boiling liquids.
- Maintain smooth boiling, either at atmospheric pressure or under vacuum
- Chips also prevent oxidation or decomposition
Particularly recommended for fractional distillations, evaporations, extractions, refluxings, condensations, and other boiling operations.
: Supplied in a 226.8 g (8 oz.) jar.