Sodium chloride ≥99.0% ACS, Analytical reagent (AR), Rankem®
Supplier: RANKEM
Certificates
About this item
CAS Number:7647-14-5
:
Formula:NaCl
MDL Number:MFCD00003477
:58.44 g/mol
Boiling Pt:1413 °C (1013 hPa)
Melting Pt:801 °C
Density:2,16 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-598-3
Specification Test Results
- Guarantee Analysis:.
- Assay (NaCl):Min. 99.00%
- pH (5% Aqueous Solution):5.00 --> 8.00
- Identity:Passes test
- Maximum Limits of impurities:.
- Insoluble Matter:Max. 0.005%
- Loss On Drying:Max. 0.50%
- Barium (Ba).:Passes test
- Arsenic(As):<=0.00004%
- Bromide(Br):<=0.01%
- Chlorate And Nitrate (As No3):<=0.003%
- Iodide:<=0.002%
- Ferrocyanide:<= 0.0001%
- Iron (Fe):<=0.0002%
- Phosphate (PO4).:<=0.0005%
- Sulphate (SO4):<=0.002%
- Total Nitrogen (N):<=0.001%
- Calcium (Ca):Max. 0.002%
- Heavy Metals (As Pb):Max. 0.0005%
- Magnesium (Mg):Max. 0.001%
- Potassium (K):Max. 0.005%