
Benzyl alcohol ≥99.00%, Laboratory Reagent, Rankem®
Supplier: RANKEM
Certificates
About this item
CAS Number:100-51-6
:
Formula:C₆H₅CH₂OH
MDL Number:MFCD00004599
:108.14 g/mol
Boiling Pt:205 °C (1013 hPa)
Melting Pt:–15,3 °C
Density:1,05 g/cm³ (20 °C)
Flash Pt:101 °C
EINECS:202-859-9
:GHS07
Specification Test Results
- Identity (IR):Conforms
- Guarantee Analysis:.
- Assay(GC):Min. 99.00%
- Weight Per ml (at 20 °C):1.045 --> 1.046 g
- Color:≤20 APHA
- Maximum Limits Of Impurities:.
- Residue On Evaporation:Max. 0.05%
- Benzaldehyde:Max. 0.20%
- Free Acid (as C6H5COOH):Max. 0.052%