Order Entry
Singapore
ContactUsLinkComponent
Multi-Use Cytotoxic Drug Spills Kit, Guest Medical
Multi-Use Cytotoxic Drug Spills Kit, Guest Medical
Catalog # GUESH9612
Supplier:  GUEST
Multi-Use Cytotoxic Drug Spills Kit, Guest Medical
Catalog # GUESH9612
Supplier:  GUEST
Specifications
  • Description:
    Multi-Use Cytotoxic Drug Spills Kit
  • Pk:
    1 KIT
Specifications
About this item
The kit contains everything required for both liquid and powder spillages in one case that can be carried to the site where it is needed.
  • 2 shakers of CT-Zorb Granules
  • 3 Hazard Scoops and Scrapers
  • 3 disposable tweezers
  • 3 absorbent pads
  • 3 sterile saline pods
  • 6 detergent wipes
  • 3 cytotoxic waste disposal bags
  • 3 bag ties
  • 3 pairs long cuff nitrile gloves
  • 3 protective disposable gowns
  • 3 pairs overshoes
  • 3 FFP3 face masks
  • Reusable goggles (BS EN 166 conformant)
  • Warning sign
  • Full instructions for use
The kit contains inert CT-Zorb Granules which will quickly and efficiently absorb all the moisture of a liquid spillage, thus containing the spill and preventing it spreading further. The granule-spill mixture can then be easily collected using the Guest Medical Scoops and Scrapers for disposal according to local procedure or regulations. Absorbent pads provide a simple method for collecting powder spillages when used with the sterile saline pods provided in the kit.