


Cyclohexanone ≥99.00%, Laboratory Reagent, Rankem®
Catalog # RANKC0451
Supplier: RANKEM
CAS Number:
Specifications
- Pack type:Glass bottle, amber
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Assay (GC):Min. 99.00 %
- Weight Per ml (at 20 °C):0.945 --> 0.947 g
- Refractive Index (at 20 °C):1.450 --> 1.451
- Water (By KF):Max. 0.20 %
Specification Test Results
About this item
:
:98.14 g/mol
Boiling Pt:155,6 °C (1013 hPa)
Melting Pt:–31 °C
Density:0,9478 g/cm³ (20 °C)
Flash Pt:43 °C
EINECS:203-631-1
UN:1915
ADR:3,III
:GHS02
:GHS05
:GHS07
CAS Number:108-94-1
Formula:C₆H₁₀(=O)
MDL Number:MFCD00001625